Learn More
Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread
Product Code. 10369990
Change view
Click to view available options
Thread Size:
GL 28
GL 30
GL 36
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size unitSize
10369990 GL 28 Each
10024750 GL 30 Each
10496881 GL 36 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10369990 Supplier Poulten & Graf GmbH Supplier No. 101 09028
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Complete a FORTUNA™ Dispenser with an adapter that comes in a wide selection of threads. Poulten & Graf GmbH Polypropylene Adaptor for FORTUNA™ Dispenser with Thread enables smaller containers to be supported on top the existing reservior.
  • Suitable for OPTIFIX and POLYFIX dispenser
  • Made of Polypropylene
  • Adapter with threads
  • Can be used for most chemicals and liquids
Specifications
Product Type Adapters for OPTIFIX and POLYFIX Dispensers
Autoclavable Autoclavable
Color White
Description Adapters for OPTIFIX and POLYFIX dispensers
Material Polypropylene
For Use With (Equipment) OPTIFIX and POLYFIX dispensers
Quantity 1
Thread Size GL 28