Learn More
Kartell™ Polypropylene 120° 'Y' Shaped Tapered Connectors
BIOPHARMA
The ideal leak-proof solution for connecting tubes. Kartell™ 120° 'Y' Shaped Tapered Connectors are useful for straight connecting tubes of various diameters.
Brand:  Kartell™ 7053000
Product Code. 12529756
  • ONLINE EXCLUSIVE PRICE
    21.40€
    14.71€ / Pack of 10
    Save 6.69€
    31% Off
Only null left
Estimated Shipment: 20-01-2026
Add to basket
Description
Description
  • Y shaped
  • Three tips separated by 120°