Learn More
Vitlab™ Polyoxymethylene Forceps
Manufactured with POM for excellent dimensional stability. VITLab™ Polyoxymethylene Forcep have grooves on the outside for optimum handling and grip.
2.20€ - 86.50€
Specifications
Tip Style | Round |
---|---|
Material | POM |
Product Type | Forceps |
Quantity (No. per Case) | 5/Pack |
For Use With (Application) | For autoclaving, bottling and decanting |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
16543642
|
Vitlab™
68099 |
115 mm |
86.50€
Pack of 50 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
11847910
|
Vitlab™
68099 |
115 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10291632
|
Vitlab™
68099 |
115 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11837910
|
Vitlab™
68199 |
145 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- For bottling and decanting
- Yello
- Blunt
- Elastic
- Very good resilience
Specifications
Round | |
Forceps | |
For autoclaving, bottling and decanting |
POM | |
5/Pack | |
Forceps |