Learn More
Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips
Product Code. 10244141
Click to view available options
Color:
Blue
Green
Red
Turquoise
Yellow
Unit Size:
Pack of 10
Product Code. 10244141
Brand: Lenz Laborglasinstrumente™ 01103710
Only null left
Add to basket
Add to basket
Ensures a secure connection while preventing damage to glass tubing and increasing safety in the laboratory. Lenz Laborglasinstrumente™ Keck™ POM Conical Joint Clips are manufactured from temperature resistant polyoxymethylene (POM).
  • Good spring properties
  • Color coded by size
  • Will not corrode or bend out of shape
Specifications
Quantity 10/Pk.
Type Joint Clip
Material POM
For Use With (Equipment) For conical ground joints
Color Turquoise