Learn More
Macherey-Nagel™ Polygram CEL 400-10 Glass Plates
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
Unit Size:
Pack of 25
Pack of 50
Specifications
Specifications
| Quantity | 50 Pk. |
| Type | Glass Plate |
| Thickness (Metric) | 0.1 mm |
| Dimensions (L x W) | 10 x 20 cm |