Learn More
Macherey-Nagel™ Polygram Alox N/UV254 Polyester Sheets
Product Code. 10236381
Change view
Click to view available options
Dimensions (L x W):
4 x 8 cm
5 x 20 cm
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
10236381 5 x 20 cm Pack of 50
10252901 4 x 8 cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10236381 Supplier Macherey-Nagel™ Supplier No. 802022
Only null left
Add to basket
Add to basket
Macherey-Nagel Polygram Alox N/UV254 Polyester Sheets, Dimensions: 4 x 8 cm, Pore Size: 60 A, Fluorescent Indicator: UV254, Thickness: 0.2 mm
Specifications
Pore Size 60 Å
Quantity 50 Pk.
Type Polyester Sheet
Fluorescent Indicator UV254
Thickness (Metric) 0.2 mm
Dimensions (L x W) 5 x 20 cm