Learn More
Macherey-Nagel™ Polygosil™ Phase
Product Code. p-8012383
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
7 μm
10 μm
Pore Size:
100 Å
1000 Å
300 Å
60 Å
Unit Size:
100g
10g
Product Code. 10728232
Brand: Macherey-Nagel™ 711530.100