Learn More
Macherey-Nagel™ Polygoprep™ 1000 to 30 C18
Product Code. 10787842
Click to view available options
Quantity:
0.1 kg
1 kg
Particle Size:
25 to 40 μm
40 to 63 μm
Unit Size:
1000g
100g
Product Code. 10787842
Brand: Macherey-Nagel™ 711028.100
Specifications
Particle Size 25 to 40 μm
Pore Size 1000 Å
Stationary Phase POLYGOPREP 1000 to 30
pH 2 to 8
Quantity 0.1 kg
Sorbent POLYGOPREP C18
Type Polygoprep
For Use With (Application) Liquid Chromatography