Learn More
Macherey-Nagel™ Polygoprep™ 100 to 12 C18
Product Code. 10358151
Click to view available options
Quantity:
0.1 kg
1 kg
Unit Size:
1000g
100g
Product Code. 10358151
Brand: Macherey-Nagel™ 711018.100
Specifications
Particle Size 10 to 15 μm
Pore Size 100 Å
Stationary Phase POLYGOPREP 100 to 12
Technique Liquid Chromatography
pH 2 to 8
Quantity 0.1 kg
Sorbent POLYGOPREP
Surface Area (Metric) 280 m2/g
Type Polygoprep
Material Silica
For Use With (Application) Liquid Chromatography