Learn More
Masterflex™ Polyethylene Reducing Connector
Description
- Quick and easy assembly without tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
- Chemical resistant
- Available in a variety of sizes
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Polyethylene |
| Tubing Size | 3/8 in. x 1/2 in. |
| Hose Barb | 3/8 in. x 1/2 in. |
| Outlet Port | 3/8 in. |
| Port Size | 1/2 in. |
| Sterilization Method | Ethylene Oxide |
| Color | Translucent |
| Product Type | Reducing Connector |