Learn More
Masterflex™ Polyethylene Pipe Adapter Elbow Connector Barbed Fitting
Product Code. 11745509
Change view
Click to view available options
For Use With:
1/4 NPT(M) x 1/4 in. tubing I.D.
1/4 NPT(M) x 3/8 in. tubing I.D.
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
11745509 1/4 NPT(M) x 1/4 in. tubing I.D. Pack of 10
11759859 1/4 NPT(M) x 3/8 in. tubing I.D. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11745509 Supplier Masterflex™ Supplier No. 3062296
Edge
Specifications
Material Polyethylene
For Use With 1/4 NPT(M) x 1/4 in. tubing I.D.
Packaging Quantity 10 Pack
Thread Size 1/4 NPT(M)
Includes Pipe adapter Elbow connector; ETO sterilization
Product Type Barbed Fitting