Learn More
Fisherbrand™ Polyethylene Overlay Sheet
Product Code. 12378546
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-4534 Gel Dryer System
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) unitSize
12378546 Fisherbrand GD-4534 Gel Dryer System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12378546 Lieferant Fisherbrand™ Lieferanten-Nr. GDPPS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Alternative Produkte anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Accessory for use with vacuum gel dryers.
  • Detachable
  • Allows gel dryer to be adapted for thin polyacrylamide gel, thicker agarose gels
  • Replacement part for Vacuum Gel Dryer systems
  • Pore size of porous polyethylene cover sheet:  0.75mm
Spezifikation
For Use With (Equipment) Fisherbrand GD-4534 Gel Dryer System
Type Sheet