Learn More
Pal International™ Polyethylene Oversleeves
Product Code. 30158553
Change view
Click to view available options
Color:
Blue
Red
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30158553 Red Pack of 100
30158548 Blue Pack of 100
30158551 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30158553 Supplier Pal International™ Supplier No. R68530
  • ONLINE EXCLUSIVE PRICE
    4.45€
    4.40€ / Pack of 100
    Save 0.05€
    1% Off
Estimated Shipment: 11-12-2026
Offer a secure, comfortable fit. Pal International™ Polyethylene Oversleeves include concealed elastic at the wrist and elbow.
  • High quality polyethylene construction
  • Concealed elastic at the wrist and elbow
  • Secure yet comfortable fit.
  • Provides splash protection
  • Ensure hygiene standards are met in a variety of industrial, manufacturing and healthcare environments.
  • Suitable for use with food
  • Latex-free to minimise the risk of irritation.
  • Easily dispensed from the Pal Ecopak dispenser
Specifications
Product Type General Purpose Sleeve
Material Polyethylene
Color Red
Disposable Single-use
Latex Free Yes