Learn More
Buerkle™ Polyethylene Hose Couplings
Product Code. 10385512 Shop All Bürkle Products
Click to view available options
Diameter (Metric):
11 to 14 mm
13 to 16 mm
3 to 5 mm
5 to 7 mm
7 to 10 mm
9 to 12 mm
Unit Size:
Pack of 10
Product Code. 10385512
Brand: Buerkle™ 87041012
Attachable, conical hose couplings for pressureless operation.
  • Manufactured from polyethylene
  • Provided 10 per pack
  • Available for a range of hose diameters
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EU Foodstuffs, RoHS
Diameter (Metric) 9 to 12 mm
Autoclavable Autoclavable
Material Polyethylene
For Use With 9 to 12 mm Hose
For Use With (Application) Pressureless Operation
Packaging Quantity 10/Pk.
Shape Conical
Product Type Hose Coupling