Learn More
Bel-Art™ Scienceware™ Polyethylene Drying Tubes
Product Code. 11891263
Change view
Click to view available options
Fits Tubing:
6.4 to 9.5 mm
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fits Tubing Unit Size
11891263 6.4 to 9.5 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11891263 Supplier Bel-Art™ Scienceware™ Supplier No. F199600000
  • ONLINE EXCLUSIVE PRICE
    35.25€
    29.44€ / Pack of 12
    Save 5.81€
    16% Off
Estimated Shipment: 09-12-2026
Made of polyethylene. Bel Art™ Polyethylene Drying Tubes can be filled with desiccant of your choice.
  • Includes two polypropylene fittings with serrated, tapered end that accept 6.4 to 9.5mm I.D. tubing
  • Unbreakable polyethylene straight tubes; 16mm I.D. x 19mm O.D.
  • Drying agent not included
Specifications
Fits Tubing 6.4 to 9.5 mm
Material Polyethylene
Product Type Drying Tube