Learn More
Piercan™ Polyethylene Dry Box Gloves
Click to view available options
Diameter (Metric) Cuff:
156
186
300
Size:
7
8
9
Unit Size:
Pack of 2
Specifications
Specifications
| Certifications/Compliance | CE 0333 Certified |
| Color | White |
| Disposable | Single-use |
| Material | Polyethylene |
| Product Type | Dry Box Gloves |
| Size | 8 |
| Sterility | Sterile |
| Temperature (Metric) Max | 25°C |
| Thickness (English) | 15 mil |
| For Use With (Application) | Nuclear industries, Life sciences |
| Show More |