Learn More
asecos™ Polyethylene Drum Chock (3-piece package)
Product Code. 17856376 Shop All Asecos Products
Change view
Click to view available options
Width (English):
4.72 in.
6.38 in.
Unit Size:
Pack of 3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (English) Unit Size
17856376 6.38 in. Pack of 3
17826416 4.72 in. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17856376 Supplier asecos™ Supplier No. 18291
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Polyethylene Drum Chock (3-piece package) is ideal for the safe emptying of drums.
  • 3-piece package
  • PE Drum Chock
Specifications
Product Type Tap
Color Brown
Material Polyethylene
Quantity 3
Width (English) 6.38 in.
Width (Metric) 162.00 mm
Height (English) 6.14 in.
Height (Metric) 156.00 mm
Weight (English) 2.97 lb.
Weight (Metric) 1.346 kg