Learn More
SCAT Corrugated Tube for HPLC
Shop All SCAT Europe Products
Click to view available options
Diameter (Metric) Inner:
6.5 mm
Unit Size:
1m
Description
Be prepared for HPLC with SCAT™ corrugated tubing.
- Flexible tubing
- For HPLC instruments
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 6.5 mm |
| Material | HDPE |
| For Use With | HPLC Instrument |
| For Use With (Application) | Leak Connection of Various HPLC Systems |
| Color | Gray |
| Product Type | Compression Union |
Product Content Correction
Product Title
Spot an opportunity for improvement?Condividi una correzione di contenuto