Learn More
Kartell™ Plastilab™ Pipette Basket
Product Code. 11300455
Click to view available options
:
Basket for Pipet Washer
Product Code. 11300455
Brand: Kartell™ 0021900
Only null left
Add to basket
Edge
Add to basket
Specially designed basket for use with Automatic Pipette Rinser.
  • Minimizes breakage
  • Buffers tips of pipettes against soft PE base
Specifications
Description Buffers tips of pipettes against soft PE base
For Use With (Application) For pipette rinser
Type Basket