Learn More
Masterflex™ Polyethylene Barbed Tee Connector
Click to view available options
Tubing Size:
1/2 in.
5/8 in.
Unit Size:
Pack of 10
Pack of 100
Description
- Ideal for low-pressure applications
- Chemical resistant
- Available in a variety of sizes
- Economical barbed fittings easily connect fluid paths without the use of tools
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
| Description | Barb Connection |
| Autoclavable | Not Autoclavable |
| Diameter (English) Inner | 15/32 in. |
| Material | HDPE |
| Overall Length (English) | 2 31/32 in. |
| Packaging Quantity | 10 Fittings |
| Tubing Size | 5/8 in. |
| Outlet Port | 5/8 in. |
| Port Size | 5/8 in. |
| Sterilization Method | Ethylene Oxide |
| Show More |