Learn More
Bryson Packaging™ Polyethylene Bags
Product Code. 12330499
Click to view available options
Width (Metric):
100 mm
250 mm
300 mm
375 mm
Height (Metric):
150 mm
200 mm
250 mm
Quantity:
100 Pack
50 Pack
Unit Size:
Pack of 100
Pack of 50
Product Code. 12330499
Brand: Bryson Packaging™ BAJ300090U
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Transparent and excellent for shipping, storage and as sample bags.
  • Available in a wide range of sizes/capacities
  • Suitable for food use
Specifications
Description Bag
Material Polyethylene
Quantity 100 Pack
Width (Metric) 250 mm
Height (Metric) 200 mm