Learn More
Brady™ Polyester Printer Labels
Product Code. 11553924
Click to view available options
Size (Metric):
25.4 x 12.7 mm (W x L), 9.53 mm dia.
25.4 x 15.24 m (W x L)
48.26 mm x 15.24 m (W x L)
9.53 mm x 15.24 m (W x L)
Unit Size:
Each
Product Code. 11553924
Brand: Brady™ 114708
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
To identify frosted, frozen, or room temperature vials with overlap
  • Permanent acrylic adhesive
  • Opaque to cover legally protected patient information
  • Matt finish
Specifications
Autoclavable Autoclavable
Certifications/Compliance RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC, UL Recognized, CSA Accepted
Description BMP71 Label Printer Labels
Adhesive Type Permanent Acrylic
Color Clear
Finish Gloss
Material Polyester
Print Technology Thermal Transfer
Product Type Printer Label
For Use With (Application) Laboratory identification, general and industrial identification, rating and nameplate labels, voice and data communication
For Use With (Equipment) BMP™71
Labels per Row 1
Quantity 1 Ea.
Size (Metric) 25.4 x 15.24 m (W x L)
Temperature Range (Metric) -70°C to +100°C
Thickness (Metric) 0.019 mm
Length (Metric) 15.24 m
Width (Metric) 25.4 mm