Learn More
Brady™ Low-profile label for use on frozen surfaces or on room-temperature surfaces.
Product Code. 15136272
Click to view available options
Color:
White
White, Transparent
Width (Metric):
9.53 mm
15.24 mm
25.4 mm
48.26 mm
Height (Metric):
9.53 mm
12.7 mm
15 mm
25.40 mm
38.1 mm
41.28 mm
44.45 mm
45.72 mm
66.68 mm
Unit Size:
Each
Product Code. 15136272
Brand: Brady™ 710779
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Low-profile label for use on frozen surfaces or on room-temperature surfaces.
  • Opaque to cover legally protected patient information
  • Ultra thin
  • Lasts in liquid nitrogen, autoclaves and hot water baths
  • Permanent Acrylic adhesive
Specifications
Certifications/Compliance 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Description BMP71 Label Printer Labels
Adhesive Type Acrylic, Permanent
Color White, Transparent
Finish Matte
Format Self-laminating
Material Polyester
Print Technology Thermal Transfer
Product Type Printer Label
For Use With (Application) Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
For Use With (Equipment) BMP™71
Labels per Row 1
Quantity 250/Pk.
Shape Rectangle
Size (Metric) 9.53W x 45.72 mmH
Temperature Range (Metric) -196°C to +110°C
Thickness (Metric) 0.012 mm
Vertical Repeat (Metric) 12.7 mm
Dimensions (LxW) 45.72 x 9.53 mm
Width (Metric) 9.53 mm
Height (Metric) 15 mm
Height (Metric) Printable Area 19.05 mm