Learn More
Brady™ Polyester Mandatory Signs
Product Code. p-8001208
Click to view available options
Displayed Text:
Hands Protection
Diameter (Metric):
200 mm
Unit Size:
Each
Product Code. 10265174
Brand: Brady™ 250111
Only null left
Specifications
Alternate Part Number PIC 255-DIA 200-B7541 (BRANDED)
Material Polyester
Quantity 1 Ea.
Adhesive Type Polyethylene, Polyester
Diameter (Metric) 200 mm
Mounting Type Self-adhesive
For Use With (Application) Outdoors Use
Max. Temperature (Metric) 120°C
Displayed Text Hands Protection
Product Type Mandatory Sign
Color White on Blue