Learn More
Brady™ Polyester Laser Radiation Warning Signs
Product Code. 11740314
Change view
Click to view available options
Height (Metric):
22 mm
43 mm
173 mm
Width (Metric):
25.00 mm
50 mm
200 mm
Unit Size:
1 roll
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) unitSize
11740314 43 mm 50 mm Each
11818030 22 mm 25.00 mm 1 roll
10244804 173 mm 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11740314 Supplier Brady™ Supplier No. 250227
Only null left
Specifications
Alternate Part Number PIC 309-TRI 050-B7541 (BRANDED)
Height (Metric) 43 mm
Width (Metric) 50 mm
Shape Triangular