Learn More
Brady™ Polyester Laser Radiation Warning Signs
Specifications
Specifications
| Alternate Part Number | PIC 309-TRI 200-B7541 (BRANDED) |
| Height (Metric) | 173 mm |
| Width (Metric) | 200 mm |
| Shape | Triangular |
| Color | Black on Yellow |