Learn More
Brady™ Polyester Labels for Thermal Transfer Printers
Product Code. 15108385 Shop All Brady Products
Change view
Click to view available options
Size (Metric):
1.27 x 4.57 cm (W x H)
1.52 x 6.67 cm (W x H)
1.9 x 4.57 cm (W x H)
1.9 x 5.59 cm (W x H)
2.54 x 1.9 cm (W x H)
2.54 x 2.54 cm
6.35 W x 6.35 cm H
Finish:
Matte
For Use With (Equipment):
BBP11, BBP12, THT 25 mm Core Printers
BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Quantity:
1500/Pk.
2500/Pk.
5000/Pk.
Unit Size:
1 roll
Pack of 2500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Finish For Use With (Equipment) Quantity Unit Size
15108385 2.54 x 1.9 cm (W x H) Matte BBP11, BBP12, THT 25 mm Core Printers 2500/Pk. 1 roll
12809948 1.52 x 6.67 cm (W x H) Matte BBP11, BBP12, THT 25 mm Core Printers 1500/Pk. 1 roll
30041436 6.35 W x 6.35 cm H Matte BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers 2500/Pk. Pack of 2500
13455748 2.54 x 1.9 cm (W x H) Matte BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers 5000/Pk. 1 roll
15348274 2.54 x 2.54 cm Matte BBP11, BBP12, THT 25 mm Core Printers 1500/Pk. 1 roll
12678293 1.9 x 5.59 cm (W x H) Matte BBP11, BBP12, THT 25 mm Core Printers 1500/Pk. 1 roll
30041400 6.35 W x 6.35 cm H Matte BBP11, BBP12, THT 25 mm Core Printers 2500/Pk. Pack of 2500
15561203 1.9 x 4.57 cm (W x H) Matte BBP11, BBP12, THT 25 mm Core Printers 1500/Pk. 1 roll
12646045 1.27 x 4.57 cm (W x H) Matte BBP11, BBP12, THT 25 mm Core Printers 1500/Pk. 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15108385 Supplier Brady™ Supplier No. 116389
  • ONLINE EXCLUSIVE PRICE
    354.00€
    298.90€ / 1 rolls
    Save 55.10€
    15% Off
View all promotions
In Stock
For barcoding and identification purposes
  • Available with gloss or matte finish
  • Available in different widths and roll lengths
Specifications
Certifications/Compliance RoHS 2002/95/EC
Description FreezerBondz Thermal Transfer Printer Labels
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Laboratory identification such as vials, centrifuge tubes, test tubes, straws, and slides
For Use With (Equipment) BBP11, BBP12, THT 25 mm Core Printers
Horizontal Repeat (Metric) 27.94 mm
Labels per Row 2
Min. Operating Temperature (Metric) -196°C
Model THT-88-492-2.5-SC
No. per Pack 2500 Label(s) / Roll
Quantity 2500/Pk.
Shape Rectangle
Size 25.40 mm (W) x 19.05 mm (H)
Size (Metric) 2.54 x 1.9 cm (W x H)
Temperature Range (English) -320°F to 230°F
Temperature Range (Metric) -196°C to 130°C
Thickness (English) 0.0032 in.
Thickness (Metric) 0.081 mm
Vertical Repeat (Metric) 22.23 mm
Dimensions (L x W) 25.40 mm (W) x 19.05 mm (H)
Length (English) 0.75 in.
Width (English) 1 in.
Width (Metric) 2.54 cm
Width (English) Printable Area 25.40 mm
Height (English) 0.75 in.
Height (Metric) 19.05 mm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Alternate Part Number THT-88-492-2.5-SC