Learn More
Brady™ GlobalMark™ Polyester Cold Temperature Tape
Product Code. 11549263
Click to view available options
Dimensions (LxW):
30.48 m x 101.6 mm
30.48 m x 107.95 mm
30.48 m x 28.58 mm
Color:
White
Yellow
Unit Size:
Each
Product Code. 11549263
Brand: Brady™ 076691
GlobalMark™ Polyester Cold Temperature Tape permanently adheres to surfaces at temperatures as low as -10 degrees F. Great for use in any cold storage warehouse, cooler or freezer. Not recommended for plotter cutting.
  • GlobalMark™ cold temperature tape permanently adheres to surfaces at temperatures as low as -10°F.
  • Great for use in any cold storage warehouse, cooler or freezer.
  • Not recommended for plotter cutting.
Specifications
Certifications/Compliance Compliant with Directive 2011/65/EU and 2015/863
Description GlobalMark Cold Temperature Tape
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Gloss
Format Roll
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) General Identification, Laboratory labelling, Safety Identification, Temperature indicating labelling
For Use With (Equipment) GlobalMark Printer
Gloss 20 Gardner Units
Labels per Row 1
Life Span 1 Year
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Temperature Range (English) -40°F to +212°F
Temperature Range (Metric) -40°C to +100°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.076 mm
Dimensions (LxW) 30.48 m x 107.95 mm
Length (English) 100 ft.
Length (Metric) 30.48 m
Width (English) 4.25 in.
Width (Metric) 107.95 mm
Alternate Part Number 076691