Learn More
Ratiolab™ Polycarbonate Cryo Boxes
Click to view available options
Color:
Blue
Unit Size:
Pack of 5
Specifications
Specifications
| Color | Blue |
| Material | Polycarbonate |
| Type | Cryo Box |
| For Use With (Application) | Liquid Nitrogen |
| Holds | 81 (1.2 and 2 mL Cryo Tubes) |
| Max. Temperature (Metric) | 121°C |
| Min. Temperature (Metric) | -190°C |
| Quantity | 5 Pack |
| Length (Metric) | 132 mm |
| Width (Metric) | 132 mm |
| Show More |