Learn More
Sperian™ Polyamide Webbing Anchorage Sling
Product Code. 12662793
Change view
Click to view available options
Length (Metric):
1.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
12662793 1.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12662793 Supplier Sperian™ Supplier No. 1002919
  • ONLINE EXCLUSIVE PRICE
    30.45€
    17.15€ / Pack of 1
    Save 13.30€
    43% Off
Estimated Shipment: 15-01-2027
Specifications
Certifications/Compliance ISO 9001 / 2000
For Use With (Application) The sling allows a reliable anchorage point to ensure full safety.
Weight (Metric) 0.25kg
Length (Metric) 1.5 m
Material Polyamide
Product Type Webbing Anchorage Sling