Learn More
Thermo Scientific™ Polyamide Layer Sheets for Chromatography
Product Code. 11469010
Click to view available options
Quantity:
1/ea.
5/ea.
Unit Size:
Each
Pack of 5
Product Code. 11469010
Brand: Thermo Scientific™ A12572.KU
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Polyamide used as a catalyst in the epoxidation of cyclohexene with tert-butylhydroperoxide
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Polymer-supported reagents and catalysts
  • Easy recovery upon completion of reaction
  • Used for Chromatography
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Ambient temperatures
Material Polyamide
For Use With (Application) Chromatography
Quantity 5/ea.
Dimensions (L x W) 15 x 15cm