Learn More
Thermo Scientific™ Polyamide Layer Sheets for Chromatography
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Polymer-supported reagents and catalysts
- Easy recovery upon completion of reaction
- Used for Chromatography
Specifications
Specifications
| Content And Storage | Ambient temperatures |
| Material | Polyamide |
| For Use With (Application) | Chromatography |
| Quantity | 1/ea. |
| Dimensions (L x W) | 15 x 15cm |