Learn More
RETSCH Polyamide Grinding Balls
Brand:  RETSCH 05.368.0042
1.95 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 10064821
  • 2.05€ / Each
Estimated Shipment: 04-07-2024
Specifications
Grinding Balls
5 mm
mm 200 andmm 301
Polyamide