Learn More
Bohlender™ Polyamide Fork Wrench
Click to view available options
Color:
Black
Unit Size:
Each
Specifications
Specifications
| Product Type | Fork Wrench |
| Material | Polyamide |
| Color | Black |
| No. per Pack | 1 Ea. |
| Temperature Range (Metric) | −10° to +140°C |