Learn More
Scilabub™ Polyamide Fiber Protective Gloves
Product Code. 11883841
Click to view available options
Size:
Small
Medium
Large
Unit Size:
1 pair
Pack of 2
Product Code. 11883841
Brand: Scilabub™ GAU/NM
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These Nomex™ polyamide heat resistant gauntlet gloves are resistant to 260°C (working temperature, dry heat only) with knitted fabric construction for extra comfort.
  • Heat-resistant (260°C working temperature, dry heat only)
  • Low thermal conductivity
  • Asbestos free
  • Double thickness
  • Knitted fabric for comfort
  • Does not shrink or melt
  • Unaffected by washing
  • Hard wearing
  • Conform to EN338: 2 X 4 X and EN407: 41324X
Specifications
Certifications/Compliance CE Mark
Color Red
Material Polyamide
Powdered Powder Free
Product Type Protective Gloves
Size Medium
Sterility Non-sterile
Resistant To Heat, Chemical
Length (Metric) 52 cm