Description
- With discharge angle and rotatable spout preventing drips
- With tapered spout for bottles with narrow openings with a diameter from 6 to 8 mm
- Media-contacting parts PA conductive to electricity and PTFE
- Supplied with PA adapter conductive to electricity for 0.75 in. inner thread
- Tight-closing for complete seal
- With connecting cap nut, guaranteed vertical position of the cock
- Pressure-resistant up to 2 bar at 20°C (without discharge bracket and grommet)
Specifications
Specifications
| Type | Electroconductive Stopcock |
| Material | Polyamide |
| Length (Metric) | 150 mm |
| For Use With (Application) | Solvents |
| For Use With (Equipment) | Nearly all commercially available canisters and barrels with 0.75 in. outer threads or (with the supplied adapter) 0.75 in. inner threads |
| Includes | Polyamide adapter |
| Color | Black |