Learn More
Koflo Unmounted In-Line Static Mixers
Product Code. 11744109 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
121 mm
127 mm
152 mm
241 mm
254 mm
Unit Size:
Pack of 10
Product Code. 11744109
Brand: Koflo 0466714
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Unmounted In-Line Static Mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table.
  • They work well with flexible tubing.
Specifications
Diameter (English) 0.2 in.
Diameter (Metric) 6 mm
Length (English) 6 in.
Length (Metric) 152 mm
Type Unmounted In-line Mixer