Learn More
Koflo Mounted In-Line Static Mixers
Product Code. 11724149 Shop All Cole-Parmer Products
Click to view available options
Length (Metric):
89 mm
184 mm
286 mm
Unit Size:
Pack of 10
Product Code. 11724149
Brand: Koflo 0466816
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Mounted In-Line Static Mixers can be used with any compression fitting with the same tubing OD.
  • For use with compression fitting with the same tubing OD
Specifications
Diameter (English) 0.5 in.
Diameter (Metric) 13 mm
Length (English) 11.3 in.
Length (Metric) 286 mm
Type Mounted In-line Mixer