Learn More
Buerkle™ Polyacetal Hose Clips
Product Code. p-8003065 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
10 to 11 mm
11 to 12 mm
12 to 14 mm
13 to 15 mm
15 to 17 mm
17 to 19 mm
18 to 20 mm
20 to 22 mm
23 to 25 mm
6 to 6.5 mm
9 to 10 mm
Unit Size:
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
10382482 6 to 6.5 mm Pack of 10
10054083 9 to 10 mm Pack of 10
10591954 10 to 11 mm Pack of 10
10696453 11 to 12 mm Pack of 10
10542403 12 to 14 mm Pack of 10
10347752 13 to 15 mm Pack of 10
10275872 15 to 17 mm Pack of 10
10163042 17 to 19 mm Pack of 10
10616843 18 to 20 mm Pack of 10
10001922 20 to 22 mm Pack of 10
10033513 23 to 25 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 10382482 Supplier Buerkle™ Supplier No. 86780006
Only null left
Add to basket
Add to basket
Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.
  • Hose clips made of Polyacetal (POM)
  • Featured in varying diameters
  • POM is white, sterilizable, does not rust, permanently elastic and hygienic.
TRUSTED_SUSTAINABILITY
Specifications
Material Polyacetal
For Use With Hoses
Packaging Quantity 10 Pc./Pk.
Tubing Size 6 to 6.5 mm
Color White
Product Type Hose Clip