missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ POLR3K Recombinant Protein
Brand: Abnova™ H00051728-P01.25ug
Additional Details : Weight : 0.00010kg
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
AAH11932 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
37.62 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MLLFCPGCGNGLIVEEGQRCHRFACNTCPYVHNITRKVTNRKYPKLKEVDDVLGGAAAWENVDSTAESCPKCEHPRAYFMQLQTRSADEPMTTFYKCCNAQCGHRWRD | |
C11/C11-RNP3/My010/RPC10/RPC11/hRPC11 | |
POLR3K | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
51728 | |
POLR3K (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
POLR3K | |
Human | |
Recombinant | |
Solution |