Learn More
Thermosafe™ PolarPack™ Refrigerant Gel Packs
Product Code. 13079332
Change view
Click to view available options
Weight (English):
8 oz./Pack
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
13079332 8 oz./Pack Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13079332 Supplier Thermosafe™ Supplier No. PP8
  • ONLINE EXCLUSIVE PRICE
    61.14€
    30.30€ / Pack of 72
    Save 30.84€
    50% Off
View all promotions
Estimated Shipment: 09-12-2026
Economical and practical refrigerant pack choice for shipping temperature-sensitive material such as food products, pharmaceuticals, and more. Thermosafe™ PolarPack™ Refrigerant Gel Packs are enclosed in durable polyethylene film.
  • Reliable non-mechanical refrigeration for the safe transport of food products, pharmaceuticals, and other medical products.
  • Multi-layered exterior minimizes surface condensation which protects product from moisture damage.
  • Outer film layer assures products arrive in top condition by absorbing condensation.
  • Phase change formulas ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2.
  • Laminated, metalized film exterior provides radiant heat barrier, resists punctures.
  • Available in several sizes
  • Reusable
  • Non-toxic
TRUSTED_SUSTAINABILITY
Specifications
Quantity 72 Pk./Cs.
Weight (English) 8 oz./Pack
Dimensions (LxWxH) 6.5 x 3.5 x 0.75 in.