Learn More
Carl Zeiss™ Polarizing Filter
Product Code. 10617804
Change view
Click to view available options
Diameter (Metric):
32 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10617804 32 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10617804 Supplier Carl Zeiss™ Supplier No. 4736000000000
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
For use with Zeiss Microscopes
Compatible Microscopes:
  • Axio Lab.A1
  • Axio Lab.A1 FL-LED
  • Axio Lab.A1 MAT
  • Axio Lab.A1 Pol
  • Axio Scope.A1
  • Axio Scope.A1 Pol
  • Axiolab
  • Axioplan 2 Imaging
  • Axioplan 2 Imaging
  • Axioplan 2 Imaging E
  • Axioplan 2 Imaging for epi-fluorescence with ApoTome equipment
  • Axioplan 2 Imaging MAT
  • Axioskop 2 FS MOT
  • Axioskop 2 FS Plus
  • Axioskop 2 MAT
  • Axioskop 2 MAT mot
  • Axioskop 2 MOT Plus
  • Axioskop 2 Plus
  • Axioskop 20
  • Axioskop 40
  • Axioskop 40 A Pol
  • Axioskop 40 FL
  • Axioskop 40 Pol
  • Axioskop 50
  • Axiostar
  • Axiostar plus
  • Axiostar plus FL
Specifications
Type Polarizing Filter, 32 mm
Diameter (Metric) 32 mm