Learn More
Thermo Scientific™ PocketTip™ D.A.R.T.'S™ (Disposable Automation Research Tips)
Product Code. 11942271
Click to view available options
Volume (Metric):
100 nL
250 nL
50 nL
Sterility:
Non-sterile
Sterile
Packaging:
20 x 384 tips
20 x 96 tips
Unit Size:
Pack of 3840
Pack of 960
Product Code. 11942271
Brand: Thermo Scientific™ 5696250
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing.
Reduce reagent, labware and sample costs while improving data quality through the elimination of intermediate pipetting steps with soft in-tip mixing. Thermo Scientific™ PocketTip™ D.A.R.T.'s™ achieve lower volume transfer capability due to a combination of the internally molded capillary pocket and the unique surface sealing of the tip.
Features:
  • Allows existing Thermo Scientific™ Matrix™ automated liquid handlers to accommodate low-volume dispensing for compound screening procedures
  • Requires only minor procedural adjustments to Thermo Scientific™ PlateMate™ and Thermo Scientific™ Hydra™ DT instrumentation
  • Eliminate aqueous dilution steps that can cause precipitation by transferring nanoliter volumes directly to assay plate.
  • Control consistent accuracy and precision during the molding process
Validation and Compatibility:
  • Purity and straightness validation are performed on compatible instrumentation to ensure only the highest quality.
  • With instrument compatibility range starting from the Matrix Hydra DT, on-the-fly dose response curves or assay development can be set up with simple Hydra DT programming.
  • Higher throughput needs can be met with the Matrix PlateMate 2 x 3 and Catalyst Express or Matrix PlateMate Plus instruments.
  • Sales specialists are available to assist with instrument programming and modifications or instrument validation of any Matrix automated liquid handler.
Ordering Information:
  • Available in 96- and 384- channel format with 50, 100 and 250nL pocket sizes
  • Sterile format available for sensitive liquid handling procedures
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 250 nL
Filtered Non-filtered
Sterility Non-sterile
Format Rack
Tip Style Automation
Packaging 20 x 96 tips
Disposable Single-use
For Use With (Equipment) Thermo Scientific Matrix Hydra DT, Thermo Scientific PlateMate Plus
Product Line PocketTip
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.