Learn More
VITLAB™ PMP Griffin Beaker
Product Code. 12783399
Change view
Click to view available options
Capacity (Metric):
25 mL
100 mL
150 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12783399 25 mL Pack of 12
12703409 100 mL Pack of 12
10210891 150 mL Each
12723409 500 mL Pack of 6
12733409 1000 mL Pack of 6
10109370 1000 mL Each
10510012 2000 mL Each
10189410 3000 mL Each
10373141 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12783399 Supplier VITLAB™ Supplier No. 60603
Only null left
Add to basket
Add to basket
Make your job easier and safer while handling a wide range of liquids with these polymethylpentene beakers. VITLAB™ PMP Griffin Beakers feature an easy to read, embossed red scale.
  • Crystal clear transparent graduated beakers
  • For autoclaving, models with raised graduations are recommended
  • To preserve markings, clean at no higher than 60°C
Specifications
Color Crystal Clear
Capacity (Metric) 25 mL
Material PMP
For Use With (Application) For Volume Measurement
Height (Metric) 50 mm
Type Griffin Beaker
Autoclavable Autoclavable
Graduated Yes
Certifications/Compliance ISO 7056
Scale Red Printed Scale
Style Griffin