Learn More
VITLAB™ PMP Griffin Beaker
Click to view available options
Capacity (Metric):
25 mL
100 mL
150 mL
500 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 12
Pack of 6
Description
- Crystal clear transparent graduated beakers
- For autoclaving, models with raised graduations are recommended
- To preserve markings, clean at no higher than 60°C
Specifications
Specifications
| Color | Crystal Clear |
| Capacity (Metric) | 3000 mL |
| Material | PMP |
| For Use With (Application) | For Volume Measurement |
| Height (Metric) | 212 mm |
| Type | Griffin Beaker |
| Autoclavable | Autoclavable |
| Graduated | Yes |
| Certifications/Compliance | ISO 7056 |
| Scale | Red Printed Scale |
| Show More |