Learn More
VITLAB™ PMP Graduated Cylinders
Brand:  VITLAB™ 64604
Additional Details : Weight : 0.67000kg
Product Code. 15722106
  • 15.20€ / Pack of 2
Estimated Shipment
30-07-2020
Add to basket
Specifications
Specifications
Round
PMP
DIN 12681, ISO 6706
Class A
145 mm
0.2 mL
10 mL
±0.1 mL
Hexagonal
15 mm
Graduated Cylinder