Learn More
Emma Safety Footwear Pluvius Safety Shoes
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather / Sympatex™ construction
- Chemical resistant / Liquid repellant
- GripForce™ Duo Heat Resistant Outsole
- Sanitized
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Closure Type | Laces |
| Color | Black, Gray |
| Design | Pluvius |
| Liner Material | Hydro-Tec™ Sanitized Silver / Sympatex™ |
| Material | Leather |
| Size | 44 |
| Toe Material | Steel |
| Resistant To | Chemicals |