Learn More
Plum™ Plutect Dual Skin Cream
Product Code. 15731137
Change view
Click to view available options
Packaging:
Bag-in-box
Tube
Unit Size:
Pack of 18
Pack of 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
15731137 Tube Pack of 18
15721137 Bag-in-box Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15731137 Supplier Plum™ Supplier No. P2541
  • ONLINE EXCLUSIVE PRICE
    114.00€
    78.96€ / Pack of 18
    Save 35.04€
    30% Off
 Restricted Item
Estimated Shipment: 23-12-2026
Skin care cream for use before and during work with alternating work processes involving water-based and other substances. May also be applied as replenishing after-work skin cream.
  • Effective protection relative to water-based and other substances
  • Perfume-, colour- and paraben-free
  • Resistant to 20 out of 20 substances in Suskind test/winner of BUS test 2007
  • Quickly absorbed, eases skin cleaning
  • Vaseline and zinc oxide provides skin protection and reduces skin irritation
  • Specifications
    Product Type Skin Care Cream
    Form Cream
    Packaging Tube
    Quantity 18 Tubes
    Volume (Metric) 100 mL