Learn More
Pluronic™ F-68 Polyol, MP Biomedicals™
Product Code. 17801431 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17801431 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17801431 Supplier MP Biomedicals Supplier No. 092750016
  • ONLINE EXCLUSIVE PRICE
    33.10€
    31.50€ / 100g
    Save 1.60€
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Nonionic detergent suitable for cell culture and insect cell culture.
Pluronic F-68 Polyol is a nonionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 g