Learn More
Thermo Scientific™ PluriTest™ Assay Service
Product Code. 18745405 Shop All Thermo Scientific Products
Change view
Click to view available options
1 Sample
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18745405 1 Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18745405 Supplier Thermo Scientific™ Supplier No. A38154
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PluriTest Assay Service is a fast and cost-effective alternative to teratoma formation for verification of pluripotency in human pluripotent stem cell lines.
  • A fast and cost-effective alternative to teratoma formation for verification of pluripotency in human pluripotent stem cell lines
  • This service leverages the GeneChip PrimeView Global Gene Expression Profile Array (905400), which enables expression profiling with an emphasis on established well-annotated content
  • More than 36,000 transcripts and variants are measured per sample
The PluriTest Assay Service offers:
  • Fast data delivery – turnaround time of four weeks or less
  • Convenient sample prep – just send a frozen cell pellet
  • Quantitative results – compare profiles to reference set of >450 cell lines and tissue types
  • Peer endorsement – assay cited >570 times in the scientific literature
  • Exceptional support – a dedicated project manager for frequent project communication
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 sample
Duration 3 Weeks
Product Type Assay Service
Product Line Applied Biosystems
Field Research Use Only
Target Audience Stem Cell Researchers
Species Human